Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCVRN
Entrez Gene ID
281447
Gene Name
recoverin
Gene Aliases
RCV1
Chromosome Location
Chr.19: 29028247 - 29036254 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174165.2 NP_776590.1 2 - 3 549 87
GenBank mRNA M95858.1 - 2 - 3 515 87
X63322.1 - - 549 87

View Full Product Details