Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM38
Entrez Gene ID
531022
Gene Name
tripartite motif containing 38
Gene Aliases
-
Chromosome Location
Chr.23: 31932143 - 31947329 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034574.1 NP_001029746.1 4 - 5 723 99
GenBank mRNA BC104589.1 - 4 - 5 745 99
BT021589.1 - 4 - 5 723 99

View Full Product Details