Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCK1
Entrez Gene ID
282855
Gene Name
phosphoenolpyruvate carboxykinase 1
Gene Aliases
PPCK1
Chromosome Location
Chr.13: 58609020 - 58615146 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174737.2 NP_777162.1 8 - 9 1451 79
GenBank mRNA AY145503.1 - 8 - 9 1451 79
BC112664.1 - 8 - 9 1454 79
JQ733516.1 - 9 - 10 1400 79
JQ733517.1 - 8 - 9 1362 79
JQ733518.1 - 8 - 9 1196 79
JQ733519.1 - 7 - 8 1263 79
JQ733520.1 - 7 - 8 1051 79

View Full Product Details