Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GID8
Entrez Gene ID
505570
Gene Name
GID complex subunit 8 homolog
Gene Aliases
C13H20orf11
Chromosome Location
Chr.13: 54464733 - 54470962 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037587.2 NP_001032676.1 2 - 3 192 76
GenBank mRNA BC109762.1 - 2 - 3 193 76

View Full Product Details