Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLBP
Entrez Gene ID
444868
Gene Name
stem-loop binding protein
Gene Aliases
-
Chromosome Location
Chr.6: 116841868 - 116857400 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045880.2 NP_001039345.1 7 - 8 791 70
GenBank mRNA AY630919.1 - 2 - 3 331 70
BC110146.1 - 7 - 8 792 70

View Full Product Details