Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYM
Entrez Gene ID
505167
Gene Name
crystallin mu
Gene Aliases
-
Chromosome Location
Chr.25: 19074616 - 19098173 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045914.2 NP_001039379.1 1 - 2 259 52
GenBank mRNA BC112846.1 - 1 - 2 261 52

View Full Product Details