Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS10
Entrez Gene ID
614550
Gene Name
regulator of G protein signaling 10
Gene Aliases
-
Chromosome Location
Chr.26: 39633166 - 39674155 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046540.2 NP_001040005.1 3 - 4 314 145
GenBank mRNA BC112856.1 - 3 - 4 316 145

View Full Product Details