Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC1
Entrez Gene ID
616832
Gene Name
doublecortin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 61674610 - 62144844 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC110025.1 - 5 - 6 1019 97

View Full Product Details