Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN15
Entrez Gene ID
522371
Gene Name
tetraspanin 15
Gene Aliases
-
Chromosome Location
Chr.28: 25803083 - 25855701 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075829.1 NP_001069297.1 2 - 3 317 64
GenBank mRNA BC116110.1 - 2 - 3 317 64

View Full Product Details