Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A16
Entrez Gene ID
614380
Gene Name
solute carrier family 22 member 16
Gene Aliases
-
Chromosome Location
Chr.9: 39753068 - 39788298 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076324.2 NP_001069792.2 5 - 6 1380 72
GenBank mRNA BC118253.1 - 5 - 6 1381 72

View Full Product Details