Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLTRN
Entrez Gene ID
513188
Gene Name
collectrin, amino acid transport regulator
Gene Aliases
TMEM27
Chromosome Location
Chr.X: 127830561 - 127878349 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075603.1 NP_001069071.1 4 - 5 397 65
GenBank mRNA BC120016.1 - 4 - 5 397 65

View Full Product Details