Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EAPP
Entrez Gene ID
520143
Gene Name
E2F associated phosphoprotein
Gene Aliases
-
Chromosome Location
Chr.21: 44930834 - 44951239 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075800.1 NP_001069268.1 5 - 6 617 144
GenBank mRNA BC119855.1 - 5 - 6 617 144

View Full Product Details