Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMD2
Entrez Gene ID
525334
Gene Name
monocyte to macrophage differentiation associated 2
Gene Aliases
-
Chromosome Location
Chr.25: 39065609 - 39127926 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075870.2 NP_001069338.1 7 - 8 669 75
GenBank mRNA BC120183.1 - 7 - 8 670 75

View Full Product Details