Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF14
Entrez Gene ID
540274
Gene Name
ring finger protein 14
Gene Aliases
-
Chromosome Location
Chr.7: 53035587 - 53054123 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081540.1 NP_001075009.1 5 - 6 1106 68
GenBank mRNA BC133406.1 - 5 - 6 1106 68

View Full Product Details