Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOK1
Entrez Gene ID
514962
Gene Name
docking protein 1
Gene Aliases
-
Chromosome Location
Chr.11: 10085659 - 10088549 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024543.2 NP_001019714.2 3 - 4 482 72
GenBank mRNA BT020685.1 - 3 - 4 482 72
BT029827.1 - 3 - 4 471 72

View Full Product Details