Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIMM29
Entrez Gene ID
508067
Gene Name
translocase of inner mitochondrial membrane 29
Gene Aliases
C7H19orf52
Chromosome Location
Chr.7: 15368548 - 15370407 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099699.1 NP_001093169.1 1 - 2 120 147
GenBank mRNA BC142113.1 - 1 - 2 120 147

View Full Product Details