Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC38A7
Entrez Gene ID
513110
Gene Name
solute carrier family 38 member 7
Gene Aliases
-
Chromosome Location
Chr.18: 26423562 - 26442537 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100355.1 NP_001093825.1 7 - 8 1194 53
GenBank mRNA BT030716.1 - 7 - 8 1194 53

View Full Product Details