Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP4B1
Entrez Gene ID
512078
Gene Name
adaptor related protein complex 4 subunit beta 1
Gene Aliases
-
Chromosome Location
Chr.3: 29479352 - 29491851 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101950.1 NP_001095420.1 2 - 3 256 60
GenBank mRNA BC149162.1 - 2 - 3 256 60

View Full Product Details