Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCARA5
Entrez Gene ID
516087
Gene Name
scavenger receptor class A member 5
Gene Aliases
-
Chromosome Location
Chr.8: 10796560 - 10929735 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102499.1 NP_001095969.1 8 - 9 1672 57
GenBank mRNA BC142199.1 - 8 - 9 1672 57

View Full Product Details