Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2C19
Entrez Gene ID
510380
Gene Name
cytochrome P450, family 2, subfamily C, polypeptide 19
Gene Aliases
-
Chromosome Location
Chr.26: 16473502 - 16520973 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109792.2 NP_001103262.1 8 - 9 1312 60
GenBank mRNA AY265992.1 - 4 - 5 540 60
BC153845.1 - 8 - 9 1314 60

View Full Product Details