Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP1S
Entrez Gene ID
527829
Gene Name
microtubule associated protein 1S
Gene Aliases
-
Chromosome Location
Chr.7: 5408684 - 5440689 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102114.1 NP_001095584.1 3 - 4 352 77
GenBank mRNA BC149678.1 - 3 - 4 352 77

View Full Product Details