Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRINP3
Entrez Gene ID
531710
Gene Name
BMP/retinoic acid inducible neural specific 3
Gene Aliases
FAM5C
Chromosome Location
Chr.16: 15141936 - 15631140 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102140.2 NP_001095610.1 3 - 4 910 115
GenBank mRNA BC151303.1 - 3 - 4 911 115

View Full Product Details