Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRX1
Entrez Gene ID
539974
Gene Name
NLR family member X1
Gene Aliases
-
Chromosome Location
Chr.15: 29751274 - 29769514 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102235.1 NP_001095705.1 6 - 7 1756 54
GenBank mRNA BC151287.1 - 6 - 7 1756 54

View Full Product Details