Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
OTOS
Entrez Gene ID
617392
Gene Name
otospiralin
Gene Aliases
-
Chromosome Location
Chr.3: 119672381 - 119673754 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114525.1 NP_001107997.1 3 - 4 145 92
GenBank mRNA BC126830.1 - 3 - 4 145 92

View Full Product Details