Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAG7
Entrez Gene ID
337901
Gene Name
pregnancy-associated glycoprotein 7
Gene Aliases
-
Chromosome Location
Chr.29: 37942844 - 37951880 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109978.1 NP_001103448.1 4 - 5 451 162
NM_176618.2 NP_788791.1 4 - 5 481 162
GenBank mRNA AB099026.1 - 3 - 4 289 163
AF020509.1 - 4 - 5 481 162
BC133469.1 - 4 - 5 489 162
EU107962.1 - 4 - 5 451 162

View Full Product Details