Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IKZF2
Entrez Gene ID
786795
Gene Name
IKAROS family zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.2: 101004607 - 101190555 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC126590.1 - 6 - 6 1453 142

View Full Product Details