Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB7
Entrez Gene ID
513568
Gene Name
serpin family B member 7
Gene Aliases
-
Chromosome Location
Chr.24: 61992313 - 62015747 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205760.2 NP_001192689.1 1 - 2 165 115

View Full Product Details