Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INHBE
Entrez Gene ID
517587
Gene Name
inhibin subunit beta E
Gene Aliases
-
Chromosome Location
Chr.5: 56005635 - 56007200 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205842.1 NP_001192771.1 1 - 2 572 77

View Full Product Details