Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLVCR1
Entrez Gene ID
533317
Gene Name
FLVCR heme transporter 1
Gene Aliases
-
Chromosome Location
Chr.16: 70677122 - 70703667 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206019.1 NP_001192948.1 5 - 6 1205 109

View Full Product Details