Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H12orf66
Entrez Gene ID
535460
Gene Name
chromosome 5 C12orf66 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 49488842 - 49511314 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206082.1 NP_001193011.1 1 - 2 233 86

View Full Product Details