Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA7
Entrez Gene ID
520403
Gene Name
carbonic anhydrase 7
Gene Aliases
-
Chromosome Location
Chr.18: 34548879 - 34557920 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192522.3 NP_001179451.1 1 - 2 40 59

View Full Product Details