Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHX4
Entrez Gene ID
540797
Gene Name
LIM homeobox 4
Gene Aliases
LXH4
Chromosome Location
Chr.16: 61427758 - 61473954 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192785.3 NP_001179714.1 4 - 5 604 59
GenBank mRNA AB257754.1 - 2 - 3 248 59

View Full Product Details