Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB3D
Entrez Gene ID
100139105
Gene Name
RAB3D, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 15738325 - 15747393 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191375.1 NP_001178304.1 1 - 2 176 100

View Full Product Details