Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC4A
Entrez Gene ID
616674
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
-
Chromosome Location
Chr.5: 101616811 - 101631660 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191510.1 NP_001178439.1 4 - 5 442 93

View Full Product Details