Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUB
Entrez Gene ID
539051
Gene Name
TUB bipartite transcription factor
Gene Aliases
-
Chromosome Location
Chr.15: 44432494 - 44457939 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192160.2 NP_001179089.1 1 - 2 35 62

View Full Product Details