Product Details

SNP ID
rs76494221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149518883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTACTATAACTTGTCTCTTTCC[A/G]TTTCACAACTAGTATCCTTTCCACG
Phenotype
MIM: 607392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WWTR1 PubMed Links

Gene Details

Gene
WWTR1
Gene Name
WW domain containing transcription regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168278.1 3329 UTR 3 NP_001161750.1
NM_001168280.1 3329 UTR 3 NP_001161752.1
NM_015472.4 3329 UTR 3 NP_056287.1
XM_017006121.1 3329 UTR 3 XP_016861610.1
XM_017006122.1 3329 UTR 3 XP_016861611.1

View Full Product Details