Product Details

SNP ID
rs74315550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:83957555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGTTTTCCTTGGTTACGTTGA[C/T]TGCGAAACTGTCCTTACTATATGGG
Phenotype
MIM: 603961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA3A PubMed Links

Gene Details

Gene
SEMA3A
Gene Name
semaphorin 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006080.2 6482 Intron NP_006071.1
XM_005250110.3 6482 UTR 3 XP_005250167.1
XM_005250111.4 6482 UTR 3 XP_005250168.1
XM_006715839.3 6482 UTR 3 XP_006715902.1
XM_011515734.2 6482 UTR 3 XP_011514036.1
XM_017011673.1 6482 UTR 3 XP_016867162.1

View Full Product Details