Product Details

SNP ID
rs77376371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:100438759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGAAGAGAGATATTGTACAGG[G/T]TGTATGTTAATAAATTTCAGTTTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MSANTD3 PubMed Links

Gene Details

Gene
MSANTD3
Gene Name
Myb/SANT DNA binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198805.1 Intron NP_001185734.1
NM_001198806.1 Intron NP_001185735.1
NM_001198807.1 Intron NP_001185736.1
NM_080655.2 Intron NP_542386.1
Gene
MSANTD3-TMEFF1
Gene Name
MSANTD3-TMEFF1 readthrough
There are no transcripts associated with this gene.

View Full Product Details