Product Details

SNP ID
rs74572897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:31316798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAAACTAAAATCAAGAGCAAT[A/T]TTAATGTGGATATTAATGACAGTGA
Phenotype
MIM: 189909 MIM: 616915
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZEB1 PubMed Links

Gene Details

Gene
ZEB1
Gene Name
zinc finger E-box binding homeobox 1
There are no transcripts associated with this gene.

Gene
ZEB1-AS1
Gene Name
ZEB1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details