Product Details

SNP ID
rs78037285
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132554352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCTGGAGGGTGGCTTAGGGCA[C/T]GCAGAGTCCACAGTGCAGGTGGACA
Phenotype
MIM: 600106
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
INPP5A PubMed Links

Gene Details

Gene
INPP5A
Gene Name
inositol polyphosphate-5-phosphatase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321042.1 Intron NP_001307971.1
NM_005539.4 Intron NP_005530.3
XM_017016204.1 Intron XP_016871693.1
XM_017016205.1 Intron XP_016871694.1

View Full Product Details