Product Details

SNP ID
rs79716763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20369515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTCCTTTCTGCTGGCGCTGCT[A/G]GTGAGGCTGGTCCGCGGGGAGGTCG
Phenotype
MIM: 123805
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE3A PubMed Links

Gene Details

Gene
PDE3A
Gene Name
phosphodiesterase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000921.4 271 Silent Mutation CTA,CTG L77L NP_000912.3
NM_001244683.1 271 Intron NP_001231612.1
XM_017019420.1 271 Silent Mutation CTA,CTG L77L XP_016874909.1
XM_017019421.1 271 Intron XP_016874910.1

View Full Product Details