Product Details

SNP ID
rs78112722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27763287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCATTTATCGTGGTGGTTTGTTT[A/T]TCTAAATTCATAGCTTTTAGAATCC
Phenotype
MIM: 611995
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MANSC4 PubMed Links

Gene Details

Gene
MANSC4
Gene Name
MANSC domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146221.1 474 Missense Mutation GAA,GAT E158D NP_001139693.1
XM_006719024.3 474 Missense Mutation GAA,GAT E113D XP_006719087.1
XM_011520542.2 474 Missense Mutation GAA,GAT E131D XP_011518844.1
Gene
MRPS35
Gene Name
mitochondrial ribosomal protein S35
There are no transcripts associated with this gene.

View Full Product Details