Product Details

SNP ID
rs76247836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40281049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTGGGGAGGCTGGGTCCTTAT[A/T]CTCCAAGGTGGACCCTTCTATTTAC
Phenotype
MIM: 608110 MIM: 604114
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD63 PubMed Links

Gene Details

Gene
ANKRD63
Gene Name
ankyrin repeat domain 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190479.2 1538 UTR 3 NP_001177408.1
Gene
BUB1B-PAK6
Gene Name
BUB1B-PAK6 readthrough
There are no transcripts associated with this gene.

Gene
PAK6
Gene Name
p21 (RAC1) activated kinase 6
There are no transcripts associated with this gene.

Gene
PLCB2
Gene Name
phospholipase C beta 2
There are no transcripts associated with this gene.

View Full Product Details