Product Details

SNP ID
rs75145098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:77422276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAGGCCTGTGTACTTTTTCTTT[G/T]GGGATTGTACTTAATCTAGCCTCAG
Phenotype
MIM: 605534 MIM: 614248
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HMG20A PubMed Links

Gene Details

Gene
HMG20A
Gene Name
high mobility group 20A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304504.1 Intron NP_001291433.1
NM_001304505.1 Intron NP_001291434.1
NM_018200.3 Intron NP_060670.1
XM_011521158.2 Intron XP_011519460.1
Gene
PEAK1
Gene Name
pseudopodium enriched atypical kinase 1
There are no transcripts associated with this gene.

View Full Product Details