Product Details

SNP ID
rs79199718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7688511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGAATTGGCGTCCGCTGTTCGC[C/T]TCTCCTCCCGGGAGTCTTCTGCCTA
Phenotype
MIM: 191170 MIM: 612661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TP53 PubMed Links

Gene Details

Gene
TP53
Gene Name
tumor protein p53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000546.5 940 Intron NP_000537.3
NM_001126112.2 940 Intron NP_001119584.1
NM_001126113.2 940 Intron NP_001119585.1
NM_001126114.2 940 Intron NP_001119586.1
NM_001126115.1 940 Intron NP_001119587.1
NM_001126116.1 940 Intron NP_001119588.1
NM_001126117.1 940 Intron NP_001119589.1
NM_001126118.1 940 Intron NP_001119590.1
NM_001276695.1 940 Intron NP_001263624.1
NM_001276696.1 940 Intron NP_001263625.1
NM_001276697.1 940 Intron NP_001263626.1
NM_001276698.1 940 Intron NP_001263627.1
NM_001276699.1 940 Intron NP_001263628.1
NM_001276760.1 940 Intron NP_001263689.1
NM_001276761.1 940 Intron NP_001263690.1
Gene
WRAP53
Gene Name
WD repeat containing antisense to TP53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143990.1 940 Intron NP_001137462.1
NM_001143991.1 940 Intron NP_001137463.1
NM_001143992.1 940 UTR 5 NP_001137464.1
NM_018081.2 940 UTR 5 NP_060551.2
XM_011523952.2 940 Intron XP_011522254.1
XM_017024812.1 940 UTR 5 XP_016880301.1

View Full Product Details