Product Details

SNP ID
rs76420255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53977322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGAGGTCTGCAGAACTGGAC[A/G]GAGCTGGGCAGTTGAGGAAACTACA
Phenotype
MIM: 606900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNG8 PubMed Links

Gene Details

Gene
CACNG8
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031895.5 Intron NP_114101.4
Gene
MIR935
Gene Name
microRNA 935
There are no transcripts associated with this gene.

View Full Product Details