Product Details

SNP ID
rs74912591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56177123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCCTTCGATGGCTCCTCCCTC[C/T]GTCCCCCTGGTCCTCCTCCTCGTCC
Phenotype
MIM: 611178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALP PubMed Links

Gene Details

Gene
GALP
Gene Name
galanin like peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145546.1 97 Silent Mutation TCC,TCT S5S NP_001139018.1
NM_033106.3 97 Silent Mutation TCC,TCT S5S NP_149097.1

View Full Product Details