Product Details

SNP ID
rs79277718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36891516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAACCAGTATGAATTCCCTCATG[A/G]CGAATGAGGTCAGAGCGACTACCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF345 PubMed Links

Gene Details

Gene
ZNF345
Gene Name
zinc finger protein 345
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242472.1 1640 Intron NP_001229401.1
NM_001242474.1 1640 Intron NP_001229403.1
NM_001242475.1 1640 Intron NP_001229404.1
NM_001242476.1 1640 Intron NP_001229405.1
NM_003419.4 1640 Intron NP_003410.1
XM_011526704.2 1640 Intron XP_011525006.1
XM_017026568.1 1640 Intron XP_016882057.1
XM_017026569.1 1640 Intron XP_016882058.1
XM_017026570.1 1640 Intron XP_016882059.1
XM_017026571.1 1640 Intron XP_016882060.1
XM_017026572.1 1640 Intron XP_016882061.1
XM_017026573.1 1640 Intron XP_016882062.1
Gene
ZNF829
Gene Name
zinc finger protein 829
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037232.3 1640 Silent Mutation CGC,CGT R425R NP_001032309.2
NM_001171979.1 1640 Silent Mutation CGC,CGT R506R NP_001165450.1
XM_005258876.4 1640 Silent Mutation CGC,CGT R425R XP_005258933.1
XM_011526933.2 1640 Silent Mutation CGC,CGT R425R XP_011525235.1

View Full Product Details