Product Details

SNP ID
rs78049648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32293537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGAAAATGAGGTCCCACCACC[C/T]GAGACCATAGTATTGTGAGAATTGC
Phenotype
MIM: 609196
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIS18A PubMed Links

Gene Details

Gene
MIS18A
Gene Name
MIS18 kinetochore protein A
There are no transcripts associated with this gene.

Gene
MRAP
Gene Name
melanocortin 2 receptor accessory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285394.1 Intron NP_001272323.1
NM_178817.3 Intron NP_848932.1
NM_206898.1 Intron NP_996781.1
XM_006724028.3 Intron XP_006724091.1
XM_017028407.1 Intron XP_016883896.1

View Full Product Details