Product Details

SNP ID
rs78996351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27929283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTTGCTATACTGCCTTCCTAA[A/C]TTGACCATGACCTCCAGTATAGTTT
Phenotype
MIM: 606876
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR3199-1 PubMed Links

Gene Details

Gene
MIR3199-1
Gene Name
microRNA 3199-1
There are no transcripts associated with this gene.

Gene
MIR3199-2
Gene Name
microRNA 3199-2
There are no transcripts associated with this gene.

Gene
PITPNB
Gene Name
phosphatidylinositol transfer protein beta
There are no transcripts associated with this gene.

Gene
TTC28-AS1
Gene Name
TTC28 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details